PTFE 6-10 micron

Lieferant: Thermo Scientific

44184.30EA 580 CHF
44184.30 44184.18 44184.A1
PTFE 6-10 micron
PTFE
Formel: (CF₂CF₂)n
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00084455
CAS-Nummer: 9002-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR