Anti-Horseradish Peroxidase Goat Polyclonal Antibody

Lieferant: Immunoreagents
IMMRIR2925EA 111 CHF
IMMRIR2925
Anti-Horseradish Peroxidase Goat Polyclonal Antibody
Antikörper
  

Type: Primary
Antigen: Horseradish Peroxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR