Quantitative migroorganisms, BioBall® HighDose 10K
56049
56052
56144
56050
56055
56143
413777
56005
BMER56049EA
0
CHF
BMER56049
BMER56052
BMER56144
BMER56050
BMER56055
BMER56143
BMER413777
BMER56005
Quantitative migroorganisms, BioBall® HighDose 10K
Mikrobiologische Testsysteme
Tests zur Überprüfung von Mirkoorganismen
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented accuracy for Quantitative Microbiological Quality Control. BioBall HighDose 10K – mean of between 8000 and 12000 cfu with a standard deviation of ≤15% of the mean (unless otherwise stated). Designed for a single precise inoculation at a higher cfu level. Ideal positive control for food testing where a higher cfu inoculation is required.
- Easy to use and requires no preparation or pre-incubation
- Can be added directly to media or matrix samples
- Accredited reference material under ISO Guide 34 standard