Sealing ring for reaction vessels
Lieferant: SciLabware
BIBB296-90EA 157 CHF
BIBB296-90
Sealing ring for reaction vessels
Versiegeln Dichtringe
  • Guarantee excellent joint seal
  • Temperature resistant from −200 to +200 °C
  • Excellent chemical resistance
O-rings for flat flanges with groove.
  • Guarantee very good joint seal
  • Temperature resistant from −200 to +200 °C
  • Very good chemical resistance