Anti-LCN8 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16750BEA 405 CHF
ABGEAP16750B
Anti-LCN8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LCN8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR