Anti-DFFA Rabbit Polyclonal Antibody

Lieferant: Chemicon
CHMCAB16965EA 1150 CHF
CHMCAB16965
Anti-DFFA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR