Anti-GFP Rabbit Polyclonal Antibody

Lieferant: US Biological
151811
USBI151811EA 648 CHF
USBI151811
Anti-GFP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR