Anti-UBQLN2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIU1000-19A-BIOTEA 1200 CHF
USBIU1000-19A-BIOT
Anti-UBQLN2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: UBQLN2 (ubiquilin 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR