Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: TL2.3]

Lieferant: United States Biological
USBIT8050-11L1EA 936 CHF
USBIT8050-11L1
Anti-TLR2 Mouse Monoclonal Antibody (Biotin) [clone: TL2.3]
Antikörper
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR