Anti-NPDC1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIN5379-02DEA 1380 CHF
USBIN5379-02D
Anti-NPDC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPDC1 (neural proliferation, differentiation and control, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR