Anti-MYT1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIM9800-80EA 825 CHF
USBIM9800-80
Anti-MYT1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Dot Blot: 1:500

Type: Primary
Antigen: MYT1 (myelin transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR