Anti-MTUS2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIM2350-72EA 889 CHF
USBIM2350-72
Anti-MTUS2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: MTUS2 (microtubule associated tumor suppressor candidate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR