Anti-ALCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H119]

Lieferant: United States Biological
USBIC2548-90M6EA 996 CHF
USBIC2548-90M6
Anti-ALCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H119]
Antikörper
Recommended dilution: Flow Cytometry: 1:50
Do not freeze!

Type: Primary
Antigen: ALCAM (activated leukocyte cell adhesion molecule)
Clonality: Monoclonal
Clone: 7H119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR