Anti-AQP5 Rabbit polyclonal antibody unconjugated

Lieferant: United States Biological
USBIA3000-24EA 676 CHF
USBIA3000-24
Anti-AQP5 Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: AQP5 (aquaporin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR