Anti-KCNIP3 Goat Polyclonal Antibody

Lieferant: United States Biological
USBI167384EA 844 CHF
USBI167384
Anti-KCNIP3 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: KCNIP3 (Kv channel interacting protein 3, calsenilin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR