Anti-FBXL2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBI150399EA 1060 CHF
USBI150399
Anti-FBXL2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: FBXL2 (F-box and leucine-rich repeat protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR