Anti-INHBB Mouse Monoclonal Antibody [clone: 13j53]

Lieferant: United States Biological
USBI140802EA 669 CHF
USBI140802
Anti-INHBB Mouse Monoclonal Antibody [clone: 13j53]
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: INHBB (inhibin, beta B)
Clonality: monoclonal
Clone: 13j53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR