Anti-Protein Phosphatase 1 Regulatory Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI136357EA 889 CHF
USBI136357
Anti-Protein Phosphatase 1 Regulatory Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Protein Phosphatase 1 Regulatory
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR