Anti-ZNF232 Mouse Monoclonal Antibody [clone: 1F8]

Lieferant: United States Biological
USBI135647EA 733 CHF
USBI135647
Anti-ZNF232 Mouse Monoclonal Antibody [clone: 1F8]
Antikörper
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: ZNF232 (zinc finger protein 232)
Clonality: monoclonal
Clone: 1F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR