Anti-RPS15 Mouse Monoclonal Antibody [clone: 4H8]

Lieferant: United States Biological
USBI132807EA 733 CHF
USBI132807
Anti-RPS15 Mouse Monoclonal Antibody [clone: 4H8]
Antikörper
Type: Primary
Antigen: RPS15 (ribosomal protein S15)
Clonality: monoclonal
Clone: 4H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR