Anti-RAB2B Mouse Monoclonal Antibody [clone: 7E4]

Lieferant: United States Biological
USBI132184EA 733 CHF
USBI132184
Anti-RAB2B Mouse Monoclonal Antibody [clone: 7E4]
Antikörper
Type: Primary
Antigen: RAB2B (RAB2B, member RAS oncogene family)
Clonality: monoclonal
Clone: 70000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR