Anti-C1QTNF7 Mouse Polyclonal Antibody

Lieferant: US Biological
USBI124110EA 733 CHF
USBI124110
Anti-C1QTNF7 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C1QTNF7 (C1q and tumor necrosis factor related protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR