Anti-ZFYVE19 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBI044079-APCEA 1200 CHF
USBI044079-APC
Anti-ZFYVE19 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ZFYVE19 (zinc finger, FYVE domain containing 19)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR