Anti-UBXN6 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI043560-BIOTINEA 1200 CHF
USBI043560-BIOTIN
Anti-UBXN6 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: UBXN6 (UBX domain protein 6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR