Anti-UBA52 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI043502-BIOTINEA 1200 CHF
USBI043502-BIOTIN
Anti-UBA52 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: UBA52 (ubiquitin A-52 residue ribosomal protein fusion product 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR