Anti-TTLL13 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBI043439-APCEA 1200 CHF
USBI043439-APC
Anti-TTLL13 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: TTLL13 (tubulin tyrosine ligase-like family, member 13)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR