Anti-TRMT10A Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI040992EA 825 CHF
USBI040992
Anti-TRMT10A Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: TRMT10A (tRNA methyltransferase 10 homolog A (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR