Anti-RC3H1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBI040916-APCEA 1200 CHF
USBI040916-APC
Anti-RC3H1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: RC3H1 (ring finger and CCCH-type domains 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR