Anti-FCHO2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
USBI035586-BIOTINEA 1200 CHF
USBI035586-BIOTIN
Anti-FCHO2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: FCHO2 (FCH domain only 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR