Anti-EIF2B3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI034992-APCEA 1200 CHF
USBI034992-APC
Anti-EIF2B3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: EIF2B3 (eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human