Anti-DTX3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBI034841-HRPEA 1200 CHF
USBI034841-HRP
Anti-DTX3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: DTX3 (deltex 3, E3 ubiquitin ligase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR