Anti-CCDC144NL Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBI033302-APCEA 1200 CHF
USBI033302-APC
Anti-CCDC144NL Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CCDC144NL (coiled-coil domain containing 144 family, N-terminal like)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR