Anti-ALDH6A1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI031778EA 825 CHF
USBI031778
Anti-ALDH6A1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50

Type: Primary
Antigen: ALDH6A1 (aldehyde dehydrogenase 6 family, member A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR