Anti-RPS6KB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 164CT21.2.2]
Lieferant: United States Biological
USBI031228-PEEA 1200 CHF
USBI031228-PE
Anti-RPS6KB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 164CT21.2.2]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Do not freeze! Light sensitive
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Monoclonal
Clone: 164CT21.2.2
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse