Anti-RPS6KB2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 164CT21.2.2]
Lieferant: United States Biological
USBI031228-APEA 1200 CHF
USBI031228-AP
Anti-RPS6KB2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 164CT21.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Monoclonal
Clone: 164CT21.2.2
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse