Anti-HSPA1A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 201CT8.2.3]

Lieferant: United States Biological
USBI031037-HRPEA 1200 CHF
USBI031037-HRP
Anti-HSPA1A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 201CT8.2.3]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-2000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: Monoclonal
Clone: 201CT8.2.3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR