Anti-IgM µ Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Lieferant: Rockland Immunochemicals
ROCK710-1807EA 719 CHF
ROCK710-1807
Anti-IgM µ Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: µ-chain
Host: Goat
Isotype: IgG
Reactivity: Mouse