Anti-FITC Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK700-101-096EA 856 CHF
ROCK700-101-096
Anti-FITC Goat Polyclonal Antibody
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:2000 - 1:8000; Western Blot: 1:200 - 1:1000;

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR