Anti-IgA Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK618-101-006EA 311 CHF
ROCK618-101-006
Anti-IgA Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR