Anti-Kappa Light Chainkappa light chain Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
ROCK610-4510EA 572 CHF
ROCK610-4510
Anti-Kappa Light Chainkappa light chain Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Secondary Rabbit Anti-κ (kappa chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:1000 - 1:2000; Western Blot: 1:500 - 1:1000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR