Anti-IgG F(c) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Rockland Immunochemicals
ROCK605-4303EA 425 CHF
ROCK605-4303
Anti-IgG F(c) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Goat IgG F(c) peroxidase conjugated antibody has been assayed against 1.0 µg of Goat IgG in a standard capture ELISA using TMB as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:16,000 is suggested for this product.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR