Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
ROCK600-105-096EA 1390 CHF
ROCK600-105-096
Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Anti-Fluorescein Antibody Alkaline Phosphatase Conjugated is suitable for immunoblotting (western or dot blot), ELISA, in situ hybridization and immunochemistry as well as other phosphatase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR