Anti-Rhodamine Mouse Polyclonal Antibody (Unconjugated) [clone: 11H10]

Lieferant: Rockland Immunochemicals
ROCK200-301-246EA 586 CHF
ROCK200-301-246
Anti-Rhodamine Mouse Polyclonal Antibody (Unconjugated) [clone: 11H10]
Antikörper
This Protein A purified monoclonal antibody against rhodamine reacts with most derivative molecules and has been tested for use in ELISA. Optimal concentration in other immunoassys should be determined by the researcher.

Recommended Dilutions: ELISA: 1:30,000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:1000 - 1:10000;

Type: Tag
Antigen: Rhodamine
Clonality: Polyclonal
Clone: 11H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR