Anti-CUL3 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-A03EA 910 CHF
ROCK100-401-A03
Anti-CUL3 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human Cul3 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).  An 88.9 kDa band corresponding to human Cul3 is detected.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CUL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR