Anti-p53 (Ab-3) (Mutant) (PAb240) Mouse Monoclonal Antibody [clone: PAb240]

Lieferant: Oncogene
OP29L-100EA 994 CHF
OP29L-100
Anti-p53 (Ab-3) (Mutant) (PAb240) Mouse Monoclonal Antibody [clone: PAb240]
Antikörper
Type: Primary
Antigen: p53 (Ab-3) (Mutant) (PAb240)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR