Anti-DeltaNp73 (38C674) Mouse Monoclonal Antibody

Lieferant: Oncogene
OP181-100EA 1080 CHF
OP181-100
Anti-DeltaNp73 (38C674) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DeltaNp73 (38C674)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR