Anti-EGFR (Ab-1) (528) Mouse Monoclonal Antibody [clone: 528]

Lieferant: Oncogene
GR01L-100EA 994 CHF
GR01L-100
Anti-EGFR (Ab-1) (528) Mouse Monoclonal Antibody [clone: 528]
Antikörper
Type: Primary
Antigen: EGFR (Ab-1) (528)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR