Anti-Fibronectin (Ab-3) (FBN11) Mouse Monoclonal Antibody [clone: FBN11]

Lieferant: Oncogene
CP70-100EA 1140 CHF
CP70-100
Anti-Fibronectin (Ab-3) (FBN11) Mouse Monoclonal Antibody [clone: FBN11]
Antikörper
Type: Primary
Antigen: Fibronectin (Ab-3) (FBN11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR