Anti-Ca V 1.2 (818-835) Rabbit Polyclonal Antibody

Lieferant: Oncogene
681507-1EA 850 CHF
681507-1
Anti-Ca V 1.2 (818-835) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Ca V 1.2 (818-835)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR