Anti-Neutrophil Elastase Rabbit Polyclonal Antibody

Lieferant: Oncogene
481001-1EA 403 CHF
481001-1
Anti-Neutrophil Elastase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Neutrophil Elastase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR